Learn More
Fisherbrand™ SureOne™ Reload Empty Racks
For use with Fisherbrand SureOne universal pipet tips
£27.30 - £28.85
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11993456
|
Fisherbrand™
T1000ER-FIS |
Standard Pipette Tips (1250 μL) |
£27.30
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11983456
|
Fisherbrand™
T200ER-FIS |
Standard Pipette Tips (200 to 300 μL) |
£28.85
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11973456
|
Fisherbrand™
T10ER-FIS |
Standard Pipette Tips (10 to 20 μL) |
£28.10
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
- Inserts (not included) sit flush with rack, allowing easy multichannel pipetting
- Notched clasp designed to prop lid open for autoclave venting
- Sure stack lid rim prevents racks from sliding around when stacked and carted around a lab
- Safe lock latch allows for one-handed opening and closing ability—won't pop open when dropped
- Sure snap design securely holds insert in place, preventing it from being lifted out while pipetting
- Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Order Info
Inserts not included; must be ordered separately.
Specifications
Empty Rack |