missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), Fisherbrand™
Use NIST-traceable reference materials for the highest accuracy available. TraceableTM Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Brand: Fisherbrand 90011-13
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Traceable Conductivity Calibration Standards Certified Reference Materials (CRM) offer easy calibration with included instructions and temperature chart. For the highest accuracy available, use NIST-traceable reference materials.
Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing. Also includes step-by step calibration instructions and temperature compensation chart. Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater.Especificaciones
16 oz. | |
Clear | |
7 to 9 |
Calibration Solution | |
Conductivity Standard | |
10 μS/cm @ 25°C |
Seguridad y manipulación
Nombre del producto
Palabra de advertencia
Clasificación
Indicaciones de peligro
Consejos de prudencia
- Traceable Conductivity Standard
Palabra de advertencia
- Warning
Clasificación
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Indicaciones de peligro
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- EUH210-Safety data sheet available on request.
Consejos de prudencia
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P370+P378-In case of fire: Use to extinguish.