missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Brand: Fisherbrand 98769-20
30.54 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
Description
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
- Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing
- Also includes step-by step calibration instructions and temperature compensation chart
- Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater
- ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
Order Info
473 mL (16 oz.) Bottle
Specifications
NIST Batch Tested, Resistivity: 150000 μ℧ | |
500 mL | |
NIST Batch Tested | |
150,000 μS |
Bottle | |
Calibration Solution | |
Conductivity Standard | |
ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material |