Learn More
Traceable™ One-Shot Conductivity Standards, Fisherbrand™
Eliminates contamination with calibration within disposable container. Traceable™ One-Shot Conductivity Standards are 100% compatible with all conductivity meters.
Brand: Fisherbrand
Code : 46
Additional Details : CAS Number : 7447-40-7 Weight : 1.00000kg
Description
Eliminates contamination with calibration within disposable container. Traceable™ One-Shot Conductivity Standards are 100% compatible with all conductivity meters.
- Nominal Value: 150000 Micro (mhos/siemens)
- Certified Reference Material
- Accuracy: certificate stated uncertainty, or ±0.25 microsiemen or ±0.25%
- Includes: 6-pack 100 mL container, step-by-step calibration instructions, individual temperature compensation charts, traceability information, Traceable Certificate
Specifications
One-Shot Conductivity Standards | |
7447-40-7 | |
Conductivity Standard | |
ClK | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 |
200,000 μS/μ℧ | |
6 x 100 mL | |
Conductivity Meters and Probes | |
MFCD00011360 | |
[Cl-].[K+] | |
74.548 | |
CHEBI:32588 |