Learn More
Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer
Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor.
£103.33 - £133.00
Specifications
Bottle Fill | Glycol |
---|---|
Certifications/Compliance | ISO 17025, A2LA, NIST |
For Use With (Application) | Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators |
Height (English) | 0.75 in. |
Height (Metric) | 19 mm |
Product Code | Brand | Model | Bottle Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Bottle Size | Price | Quantity & Availability | |||||
11749745
|
Fisherbrand™
90008-87 |
Refrigerator/Freezer Thermometer with 5mL bottle | 5 mL |
£133.00
Each |
In Stock
Log in to see stock. ![]() |
|||||
11873460
|
Fisherbrand™
90008-35 |
Refrigerator/Freezer Thermometer with 10mL bottle | 10 mL |
£103.33
Each |
In Stock
Log in to see stock. ![]() |
|||||
Description
Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor. Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer includes a probe which is sealed in glycol solution and insulates sensor when refrigerator door is opened.
- Triple display simultaneously shows Min/max and current temperatures, °F/°C switchable
- Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
- 15-077-720 has a 5mL bottle designed for vaccines; 06-664-11 has a 10mL bottle
- Glass bottle filled with glycol mimics vaccine temperature and fits in trays
- Probe is sealed in glycol solution which insulates sensor when refrigerator door is opened
- Ten-foot micro-cable permits refrigerator doors to close on it
- Minimum/maximum memory monitors high and low readings overnight, on weekends, or for any time period
- Alarm is programmable in 1° increments, sounds continuously when temperature rises above/falls below set points and can be turned off manually
- High-impact, chemical-resistant ABS plastic case
- Temperature range: -50 to +70°C/-58 to +158°F
- Resolution: 1° accuracy: ±1C
- Battery:AA Alkaline (1)
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It provides traceability to standards provided by NIST.
Specifications
Glycol | |
Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators | |
19 mm | |
70 mm | |
ABS Plastic | |
4 oz. | |
2.75 in. | |
70°C | |
-50°C | |
-50°C to +70°C | |
Refrigerators/Freezers | |
±1°C |
ISO 17025, A2LA, NIST | |
0.75 in. | |
10 in. | |
25.4 cm | |
1°C | |
Fridge/Freezer Digital Thermometer | |
158°F | |
-58°F | |
-58°F to +158°F | |
Yes | |
AA Alkaline (1) | |
Digital LCD |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.