missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Vortexer Mixer
The Vortexer is a multi-purpose and maximum convenience unit. This sleek and modern designed vortex mixer is capable of meeting the mixing requirements of the most popular test tubes used in today’s lab without the need to swap out mixing head units.
Brand: Fisherbrand™ 120904
197.45 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Body made from durable ABS to resist most commonly used chemicals, and is easy to clean
- Infinity adjustable speed rotary dial provides your choice of speed control
- Stable elastomeric device feet, provide vibration dampening and prevent movement during operation
- Innovative head can be removed for cleaning purposes
- US Design Patent No. D787,085, US Utility Patent No. 9895670
Head Piece Capacity:
- Center pad area for hand held mixing
- Assay plate x 1 (to be used with retention cords)
- 25/50 mL conical tubes x 2 (horizontally mounted)
- 15 mL conical tubes x 2 (horizontally mounted)
- 5 mL tubes x 2 (horizontally mounted in the same retention space as 15 mL)
- 1.5/2 mL microfuge tubes x 4 (vertically mounted)
- 0.5 mL microfuge tubes x 6 (vertically mounted)
- 0.2 mL microfuge tubes (strip tubes) x 24 (vertically mounted)
Specifications
3.7 mm | |
0 to 3000 rpm | |
2°C to 40°C | |
Vortex Mixer | |
CE | |
ABS plastic | |
7.8 in. | |
6.7 in. | |
7.5 in. | |
11 lb. | |
11 lb. | |
Blue | |
Multiple Tube Sizes, Assay Plates, and Vessels | |
intermittent, off, continuous |
50 mL Conical Tubes | |
220/240 V | |
74 W | |
CE, CSA, RoHS3, UKCA, WEEE | |
50/60 Hz | |
6.7 x 7.5 x 7.8 in. (17.1 x 19 x 19.7 cm) | |
19.7 cm | |
17.1 cm | |
19 cm | |
5 kg | |
5 kg | |
Vortexer Mixer, Blue | |
Vortexer unit, multi-head, central pad (3 Pk.), retention cords (3 Pk.), 220 V-240 V CE cord | |
5 Year Warranty |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Vortexer Mixer > Product Type: Vortex Mixer
Spot an opportunity for improvement?Share a Content Correction