Learn More
Fisherbrand™ Weighing Funnels
Weighing dish and sample transfer funnel in one.
Brand: Fisherbrand™
Additional Details : Weight : 0.10000kg
Description
Weighing dish and sample transfer funnel in one.
- Borosilicate glass funnels with tapered pouring necks aid microtransfer of solids and liquids
- No rocking or moving while weighing—flat bottom keeps funnel steady while on balance
- Use in temperatures to 454°C (849°F)
Order Info
Assortment pack of five includes one each of the following (diameter x L): 10 x 35mm, 15 x 40mm, 20 x 45mm, 25 x 55mm and 32 x 68mm
Specifications
25mm | |
Borosilicate Glass | |
Used for microtransfer of solids and liquids | |
454°C |
10.25mm (OD); 8.12mm (ID) | |
55 mm | |
849°F | |
Weighing Funnel |