Learn More
Fisherbrand™ Zipper Seal Sample Bags
Zipper-locking feature securely seals bags
£10.23 - £15.35
Product Code | Brand | Dimensions (L x W) | Thickness (English) | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Thickness (English) | Material | Price | Quantity & Availability | ||||
13554530
|
Fisherbrand™
018161A |
3 x 2 in. (7 x 5cm) | 2 mil | LDPE |
£10.23
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||
10613771
|
Fisherbrand™
01816D |
8 x 5 in. (20 x 13cm) | 4 mil | LDPE |
£15.35
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||
Description
Zipper-locking feature securely seals bags
- Durable, transparent polyethylene
- Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
- Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
Zip Seal |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Zipper Seal Sample Bags