missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FitBag™ Buffered Peptone Water (BPW)

Shorten your workflow with easy to prepare 2.7L, 4.5L or 9L media bags: ready in 5 to 10 minutes, while segmented structure allows pre-filling.

Brand:  Thermo Scientific™ DF0509A

377.32 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15775463

  • £357.00 / Pack of 30
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Save time in preparing selective enrichment medium for the isolation of Salmonella spp. from food samples using Thermo Scientific™ FitBag™ Buffered Peptone Water. The irradiated medium is supplied in lightweight, transparent, plastic bags—just add water and the medium is ready for use. The bag's segmented structure allows pre-filling so that you can reconstitute in two minutes, when needed. Available in 2.7L, 4.5L or 9L sizes to suit your workflow, medium can be dispensed through the outlet tube as required.

Shorten your workflow with FitBag UVM1 Broth. FitBag Enrichment Media is quick, easy-to-prepare, and highly adaptable to laboratory workflows and automated equipment, such as dilutors. No further supplementation is required.

Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.

Flexible: segmented structure allows advanced preparation by filling one segment with deionized water from which FitBag Enrichment Media can be reconstituted within two minutes. Pre-sterilized water may also be added under aseptic conditions.

Easy-to-use: fill and dispense with minimum training.

Energy saving:
gamma irradiated, eliminating the need for costly thermal processing.

Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste.

  • Reduces waste: unprepared FitBag Enrichment Media can be kept for up to 24 months. Rehydrated media has a shelf life of 10 days.

Quality guaranteed: compliant with ISO 11133 and ISO 7218. Quality certificates are available for every batch, reducing the QC testing needed.

Space-saving:
conveniently packed in flat boxes, FitBag Enrichment Media takes up relatively little space compared to prepared media and can be made up as required.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10°C to 30°C
Convenience Format
Pre-enrichment Medium
30 x 2.7 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.