missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ FITC Protein Blocking Agent (PBA)
Block nonspecific antibody binding of primary reagent with Epredia™ FITC Protein Blocking Agent (PBA)
Brand: Epredia™ TA-006-PBA
This item is not returnable.
View return policy
Description
- For In Vitro Diagnostic use
- Blocks nonspecific antibody (protein) binding of primary reagent
Storage Conditions: Store at 2°-8°C.
Recommended for: Fluorescence Microscopy.

Specifications
6 mL | |
2° to 8°C. | |
6 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ FITC Protein Blocking Agent (PBA) > 6mL
Spot an opportunity for improvement?Share a Content Correction