Learn More
SCAT™ Fittings for HPLC Systems
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
Brand: SCAT™ 160499
Code : NEW
Additional Details : Weight : 0.01000kg
Description
SCAT fittings are available in the following versions:
- PFA fitting with integrated ferrule in various colors and diameters
- Tube connectors in various diameters
Specifications
Installation Wrench | |
Solvent Handling | |
Polypropylene Fittings (e.g. Earlier Generation of Safety Caps) |
Black | |
Square Installation Wrench | |
PFA |