Learn More
SCAT™ Fittings for HPLC Systems
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
Brand:  SCAT™ 107066
 View more versions of this product
Product Code. 10715572
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
SCAT fittings are available in the following versions:
  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters
Specifications
Specifications
Fitting
3.2 mm
Solvent Handling
Fitting with Integrated Ferrule (3.2 mm OD)
PFA
Colorless
3.2 mm
5 Fittings
Safety Caps, Safety Waste Caps, HPLC and UHPLC Systems