missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Fittings for HPLC Systems

Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.

Brand:  SCAT™ 107813

8.86 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10891341

  • £11.25 / Each
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

SCAT fittings are available in the following versions:

  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters
Specifications

Specifications

Connector
4 to 6 mm
Straight Tube Connector (4 to 6 mm ID)
Polypropylene
White
Solvent Handling
4 to 6 mm ID Tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers