Learn More
Restek™ Fittings, Parker (Treated): Plugs
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
Brand:  Restek™ 22879
Encompass_Preferred
 View more versions of this product
Product Code. 13127292
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Available in Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Specifications
Specifications
Plug
1/4 in.
Silcosteel-CR Coated