Learn More
Mettler Toledo™ FiveEasy™ F20 pH/mV Meters
Compact, benchtop meter provides high quality pH/mV measurements with simple click of a button
Brand: Mettler Toledo™ 30266626
468.75 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Includes: LE438 sensor, installation QuickGuide, pH buffer starter pack
Description
- Compact, space saving design with ABS housing
- Remove sensor holder after use and store on the side of the meter
- Large, well-structured 4.3 in. segmented LCD display for easy viewing
- Intuitive button arrangement and simple menu for easy operation
- ATC and MTC temperature compensation
- Three point (max.) linear calibration with four predefined buffer groups
- BNC, Cinch, NTC 30kiloOhm sensor inputs
- Features analog output, Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
- Ideal for wide range of laboratory applications
- pH Range: 0.00 to 14.00
- pH Resolution: 0.01
- pH Accuracy: ±:0.01
- mV Range: 2000 to 2000mv
- mV Resolution: 1mV
- mV Accuracy: ±1mV
- Temperature Range: 0° to +100°C (32° to 212°F)
- Temperature Resolution: 0.1°C
- Temperature Accuracy: 0.5°C
- Memory Size: current calibration
- Power Supply: 100 to 240V, 50/60Hz, 12 VDC
- Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
- Weight: 0.63kg (1.4 lb.)
- F20 Meter only: Suited for applications that require a specific sensor not included in a kit
- F20 Standard Kit: Suited for wide range of general aqueous samples

Specifications
FiveEasy F20-Std | |
0.5°C | |
±0.01 | |
Linear | |
0.01 | |
1mV | |
32°F to 212°F | |
LCD | |
BNC, Cinch, NTC 30kiloOhm sensors | |
0.63 kg | |
ATC and MTC | |
120/240 V |
F20 pH meter, LE438 pH sensor, electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, and 10.01, test certificate, pH measurement guide | |
±1 | |
Wide range of general aqueous samples | |
0 to 14 | |
2000 to 2000 | |
0°C to 100°C | |
0.1°C | |
Current calibration | |
22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.) | |
1.4 lb. | |
F20 Standard Kit | |
50/60 Hz |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
- Multiple Manufacturers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.