missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ FiveEasy™ Plus FP20 pH/mV Meters
Compact, benchtop meter provides high quality pH/mV measurements with simple click of a button.
£660.00 - £885.00
Specifications
Accuracy (mV) | ±1 |
---|---|
Accuracy (pH) | ±0.01 |
Accuracy (Temperature) | 0.3°C |
Data Management Details | RS232, USB |
Calibration | Linear/Segmented |
Product Code | Brand | Type | Includes | For Use With (Application) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | For Use With (Application) | Price | Quantity & Availability | |||
15583370
|
Mettler Toledo™
30266940 |
FP20 Micro Kit | FP20 pH meter, LE422 pH sensor, electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, and 10.01, test certificate, pH measurement guide | For small samples in vials and test tubes |
£885.00
1 unit |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||
15563360
|
Mettler Toledo™
30266628 |
FP20 Standard Kit | FP20 pH meter, LE438 pH sensor, 2 buffer sachets each for pH 4.01, 7, and 10.01, test certificate, pH measurement guide, electrode holder, | Wide range of general aqueous samples |
£877.20
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
15593370
|
Mettler Toledo™
30266941 |
FP20 TRIS Kit | FP20 pH meter, LE420 pH sensor, electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, and 10.01, test certificate, pH measurement guide | Suitable for wide range of laboratory applications |
£820.00
1 unit |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
15573360
|
Mettler Toledo™
30266629 |
FP20 Bio Kit | FP20 pH meter, LE410 pH sensor, electrode holder, 2 buffer sachets each for pH 4.01, 7, and 10.01, test certificate, pH measurement guide | Fast results in aqueous samples such as acids, bases |
£840.00
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
15553360
|
Mettler Toledo™
30266627 |
FP20 Meter Only | FP20 pH meter, charger, test certificate, pH measurement guide | Applications that require a specific sensor not included in a kit |
£660.00
1 unit |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
Description
- Compact, space saving design with ABS housing
- Remove sensor holder after use and store on the side of the meter
- Large, well-structured 4.3 in. segmented LCD display for easy viewing
- Intuitive button arrangement and simple menu for easy operation
- RS232 or USB ports to export data directly either to a printer or a PC for further processing
- ATC and MTC temperature compensation
- Five point (max.) linear only calibration with four predefined buffer groups
- BNC, Cinch, NTC 30kiloOhm sensor inputs
- Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
- Ideal for wide range of laboratory applications
- pH Range: -2.00 to 16.00
- pH Resolution: 0.01/0.1
- pH Accuracy: ±:0.01
- mV Range: 2000 to 2000mv
- mV Resolution: 1mV
- mV Accuracy: ±1mV
- Temperature Range: -5° to +105°C (23° to 221°F)
- Temperature Resolution: 0.1°C
- Temperature Accuracy: 0.3°C
- Memory Size: 200 measurements, current calibration
- Power Supply: 100 to 240V, 50/60Hz, 12 VDC
- Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
- Weight: 0.63kg (1.4 lb.)
- FP20 Meter only: Suited for applications that require a specific sensor not included in a kit
- FP20 Standard Kit: Suited for wide range of general aqueous samples
- FP20 Bio Kit: Suited for fast results in aqueous samples such as acids, bases
- FP20 Micro Kit: Suited for small samples in vials and test tubes
- FP20 TRIS Kit: Suited for demanding, dirty samples such as emulsions, oily samples, waste water, TRIS
Specifications
±1 | |
0.3°C | |
Linear/Segmented | |
0.01, 0.1 | |
1mV | |
23°F to 221°F | |
LCD | |
BNC, Cinch, NTC 30kiloOhm sensors | |
0.63 kg | |
ATC and MTC | |
50/60 Hz |
±0.01 | |
RS232, USB | |
-2.00 to 16.00 | |
2000 to 2000 | |
-5°C to +105°C | |
0.1°C | |
200 measurements, current calibration | |
22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.) | |
1.4 lb. | |
120/240 V |