missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveGo™ F2 pH/mV Meters

Portable meter provides reliable and high quality pH/mV measurements with simple click of a button

Brand:  Mettler Toledo™ 30266946

272.70 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 15543380

  • £366.00 / 1 unit
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket

Includes: Electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack
±1
0 to 14
1mV
0.1°
200 measurements, current calibration
18.8 x 7.7 x 3.3 cm (97.4 x 3 x 1.3 in.)
0.5 lb.
F2 Meter Only
±0.5°C
Applications that require a specific sensor not included in a kit
0.01
3
LCD
BNC, Cinch
0.26 kg
ATC and MTC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ FiveGo™ F2 pH/mV Meters > F2 pH/mV meter only

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.