missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveGo™ F2 pH/mV Meters

Product Code. p-7070339 Shop All Mettler Toledo Products
Click to view available options
Type:
Meter with Field Kit
Meter with Food Kit
Meter with Standard Kit
pH/mV Meter
Unit Size:
1 unit
This item is not returnable. View return policy

Product Code. 15543380

Brand: Mettler Toledo™ 30266946

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack

Portable meter provides reliable and high quality pH/mV measurements with simple click of a button

Different combinations of portable meters, sensors and accessories are available, which are ideally suited for specific applications or samples.

  • Robust, IP67 waterproof ABS housing
  • Large, well-structured 3.1 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • Ergonomic design provide comfort to both big and small hands
  • Foldable foot stand provides optimal viewing while situated on a lab bench
  • Withstands wet and demanding environments
  • Includes compact carrying case
  • Three point calibration with four predefined buffer groups
  • ATC and MTC temperature compensation
  • Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • BNC, Cinch sensor inputs
  • Ideal for mobile applications in the laboratory, at-line, or outdoors
Specifications:
  • pH Range: 0.00 to 14.00
  • pH Resolution: 0.01
  • pH Accuracy: ±0.01mV
  • mV Range: -1999 to 1999mV
  • mV Resolution: 1mV
  • mV Accuracy: ±1mV
  • Temperature Range: 0° to +100°C (32° to 212°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: ±0.5°C
  • Memory Size: 200 measurements, current calibration
  • Operating Hours: >200 hrs.
  • Power Supply: 4 x 1.5V AAA LRO3 or 4 x 1.2V AAA NiMH HR03
  • Dimensions: 18.8 x 7.7 x 3.3cm (97.4 x 3 x 1.3 in.)
  • Weight: 0.26kg (0.5 lb.)
Applications:
  • F2-Meter only: Suitable for applications that require a specific sensor not included in a kit
  • F2 Food Kit: Suitable for solid and semi-solid samples such as meat, fruit, cheese, and more
  • F2 Field Kit: Suitable for wide range of general aqueous samples and for measurements on the go
  • F2 Standard Kit: Suitable for wide range of general aqueous samples including emulsions and suspensions
TRUSTED_SUSTAINABILITY

Specifications

Includes Electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±1
For Use With (Application) Applications that require a specific sensor not included in a kit
Measurement Modes pH
Calibration Linear
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) -1999 to 1999
Resolution (mV) 1mV
Calibration Points 3
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°
Display Type LCD
Backlight Option None
Memory 200 measurements, current calibration
Data Points 200
Log Function Type Automatic, Manual
Inputs BNC, Cinch
Dimensions (L x W x H) 18.8 x 7.7 x 3.3 cm (97.4 x 3 x 1.3 in.)
Weight (Metric) 0.26 kg
Weight (English) 0.5 lb.
Model F2
Temperature Compensation ATC and MTC
Type pH/mV Meter
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.