Learn More
Invitrogen™ FIX & PERM™ Cell Permeabilization Kit
Product Code. 10324023
Click to view available options
No. of Tests:
200 Tests
50 Tests
Unit Size:
Each
For Research Use Only. All usage must comply with product instructions.
Product Code. 10324023
Brand: Invitrogen™ GAS003
Edge
For Research Use Only. All usage must comply with product instructions.
FIX & PERM™ Cell Permeabilization Kit
The FIX & PERM™ Cell Permeabilization Kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.
The FIX & PERM™ Cell Permeabilization Kit is:
  • Flexible—compatible with multiple sample types and cellular antigens
  • Robust—scatter pattern of cells unaffected
  • Efficient—simultaneous detection of intracellular and surface markers
    View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).
    The FIX & PERM™ formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.The FIX & PERM™ Cell Permeabilization Kit is suitable for various biological samples and has been validated for use with cell lines, whole blood, and bone marrow. The FIX & PERM™ kit has been shown to accurately identify previously undetectable intracellular markers. Unlike other fixation and permeabilization reagents available on the market, the FIX & PERM™ staining protocol leaves the morphological scatter characteristics of the cells intact.
    How It Works
    The FIX & PERM™ Cell Permeabilization Kit contains 2 reagents: Fixation Medium (Medium A) and Permeabilization Medium (Medium B). Cells are incubated for 15 minutes at room temperature with Fixation Medium, washed, and then incubated with the Permeabilization Medium and fluorochrome-labeled antibody. Fixation Medium and Permeabilization Medium are also available separately (Cat. Nos. GAS001S100 & GAS002S100).
  • TRUSTED_SUSTAINABILITY
    Specifications
    No. of Tests 50 Tests
    Product Line FIX & PERM
    Quantity 50 tests
    Shipping Condition Room Temperature
    Product Type Fixation/Permeabilization Reagent