missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 780

A viability dye that can be used to irreversibly label dead cells prior to cryopreservation, fixation, and/or permeabilization procedures.

Brand:  Invitrogen™ 65-0865-18

237.64 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 15383562

  • £288.00 / 500 tests

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Unlike 7-AAD and propidium iodide, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized, and stained for intracellular antigens without any loss of staining intensity of the dead cells
  • Thus, using Fixable Viability Dyes allows dead cells to be excluded from analysis when intracellular targets are being studied
  • Fixable Viability Dye eFluor™ 780 can be excited by the red (633 nm) laser line and has a peak emission of 780 nm that can be detected using a 780/60 band pass filter (equivalent to APC-eFluor™ 780 or APC-Alexa Fluor™ 750)
  • For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
  • If the percentage of dead cells is expected to be less than 5%, then it is recommended to take a small aliquot of cells and heat them at 65°C for 1 minute then immediately place on ice for 1 minute
  • After this treatment, the heat-killed cells can be combined 1:1 with live cells and then stained with the Fixable Viability Dye
  • Testing suggests that compensation out of most detectors is negligible, with compensation out of PE-Cyanine7 being the highest at less than 5%
  • Actual compensation values will depend on each investigator's specific instrument, filter sets, and PMT voltage settings
  • Fixable Viability Dye eFluor™ 780 is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO
  • It should be protected from light and moisture
  • Store at -70°C with dessicant, it may be freeze-thawed up to 20 times
  • Allow vial to equilibrate to room temperature before opening
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Infrared
Viability Assay
Dye
eBioscience™
Fluorescence
Flow Cytometer
633/780
500 Tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.