missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fixative-Free Lysing Solution, High-Yield Lyse

Product Code. 10682074
Click to view available options
Quantity:
500 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10682074

Brand: Invitrogen™ HYL250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Fixative-Free Lysing Solution, High-Yield Lyse

High-Yield Lyse is a premixed, fixative-free erythrocyte lysing solution that can be used to eliminate red cells from whole blood for flow cytometric analysis with minimal loss of rare blood cell populations. Using this reagent, lysis of erythrocytes is performed immediately following staining of the blood samples with Invitrogen's fluorochrome-conjugated monoclonal antibodies.

View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads™ for cell isolation.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.

Shelf life: 24 months from date of manufacture
Sample Type Blood
Product Type Lysing Solution
Quantity 500 mL
Reagent Type Whole Blood Lysis Buffer
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.