Learn More
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid
For use with Centrifuge 5430 and Centrifuge 5430 R
Brand: Eppendorf™ 5427739009
861.48 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : A0
Additional Details : Weight : 4.40100kg
Description
- Aerosol-tight Eppendorf QuickLock rotors close with only 1/4 of a turn
- Saves time during every day use
- Takes repetitive stress off your wrist
Specifications
Model F-35-6-30 | |
6 x 15/50 mL | |
Fixed | |
6 |
Fixed Angle Rotor | |
5430/5430R Centrifuges | |
Lid |