Learn More
Trajan™ Fixed-Needle Autosampler Syringes for Thermo Scientific Instruments
For reduced non specific sample interaction and increased analyte recovery
Brand:  Trajan™ 0029671
 View more versions of this product
Product Code. 15208463
  • £78.50 / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Specifications
25 G
0.5 mm
1.96 in.
10F-BT-5/0.5C
10 μL
Thermo Scientific instruments AS3000, AS2000, AS200/800
0.019 in.
Gas chromatography
Autosampler Syringe
Ea.
Cone
Fixed