Learn More
Thermo Scientific™ Fixed Needle Syringes for GC Instruments
Get accurate, reproducible results from these fixed-needle syringes for Thermo Scientific™ and Agilent GC autosamplers.
£67.70 - £348.00
Product Code | Brand | For Use With (Equipment) | Volume (Metric) | Needle Gauge | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Volume (Metric) | Needle Gauge | Price | Quantity & Availability | |||
10512265
|
Thermo Scientific™
36500505 |
AS3000, AS2000, AI/AS1310, AI/AS 1610 Autosamplers | 5 μL | 26 G |
£113.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10374653
|
Thermo Scientific™
36500525 |
TriPlus, AS3000, AS2000, AS200, AS800, AI/AS 1610 Autosampler | 10 μL | 25 G |
£84.62
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10197854
|
Thermo Scientific™
36500580 |
TriPlus, AS2000, AI/AS 1610 Autosamplers | 10 μL | 23 to 26 G |
£83.75
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10384653
|
Thermo Scientific™
36502019 |
TriPlus, AS2000 Autosampler | 10 μL | 26 G |
£76.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10527694
|
Thermo Scientific™
36504047 |
TriPlus Autosampler | 5 μL | 26 G |
£75.28
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10746173
|
Thermo Scientific™
36520060 |
AI/AS1310, TriPlus, AS300, AS2000, AS200/AS800, AI/AS 1610 Autosampler | 10 μL | 23 G |
£71.80
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10721724
|
Thermo Scientific™
36520061 |
TriPlus, AS2000 Autosampler | 10 μL | 23 G |
£99.18
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10364083
|
Thermo Scientific™
365C3701 |
CTC GC Instruments, AI/AS 1610 Autosamplers | 5 μL | 23 G |
£75.83
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10029192
|
Thermo Scientific™
365D1856 |
CTC GC Instruments, AI/AS 1610 Autosamplers | 10 μL | 26 G |
£348.00
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10206824
|
Thermo Scientific™
365D3711 |
CTC GC Instruments, AI/AS 1610 Autosamplers | 10 μL | 26 G |
£67.70
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10361505
|
Thermo Scientific™
36504045 |
AI/AS1310, TriPlus, AS3000, AI/AS 1610 Autosamplers | 0.5 μL | 23 G |
£120.14
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10025744
|
Thermo Scientific™
365D2971 |
CTC GC Instruments, AI/AS 1610 Autosamplers | 10 μL | 23 G |
£259.00
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13456806
|
Thermo Scientific™
365C0241 |
TriPlus RSH Autosampler | 5 μL | 26s G |
£122.65
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Get accurate, reproducible fixed-needle syringes for Thermo Scientific™ and Agilent GC autosamplers. Choose from a variety of sizes, types and autosampler compatibilities. They are available in standard industry size with white scale backing for easy reading and solvent-resistant markings.
- Borosilicate glass with metal flanges
- Needles and plungers are stainless steel
- Field-proven syringe design
- Cone tip
Specifications
No | |
Cone |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.