Learn More
Microspec™ Fixed Volume Pipette
Single channel pipette with fixed volume
Brand: Microspec™ AD8
Additional Details : Weight : 0.06800kg
Description
- Fixed volume
- Single channel
- Non-sterile
- Grey
Specifications
0.01 | |
Gray | |
Non-sterile | |
250 μL |
Not Autoclavable | |
1 | |
Fixed Volume Pipette |