missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies FK506
Calcineurin/NFAT pathway inhibitor; Binds FKBP-12
£170.10 - £697.95
Specifications
CAS | 104987-11-3 |
---|---|
Synonym | Tacrolimus |
Molecular Formula | C44H69NO12 |
Purity | ≥99% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17180332
|
STEMCELL Technologies
74152 |
10 mg |
£170.10
10mg |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17190332
|
STEMCELL Technologies
74154 |
50 mg |
£697.95
50mg |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- FK506 is a macrolide antibiotic with potent immunosuppressive properties (Thomson et al.) It inhibits the calcineurin/NFAT pathway (Martinez-Martinez & Redondo) and binds with high affinity to FK506 binding protein 12 (FKBP-12) (Thomson et al.)
- Promotes the self-renewal of mouse embryonic stem cells independent of leukemia inhibitory factor (LIF), and attenuates differentiation following LIF withdrawal (Li et al.)
- In combination with 1-Azakenpaullone (Catalog #74252) and ID-8 (Catalog #72502), enhances the expansion rates of several human pluripotent stem cell lines (Yasuda et al.)
- Inhibits pro-inflammatory cytokine production by activated T cells (Sakuma et al.)
Specifications
104987-11-3 | |
Tacrolimus | |
C44H69NO12 | |
≥99% |