Learn More
FKBP10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562336
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CSLLDGTQLF TSHDYGAPQE ATLGANKVIE GLDTGLQGMC VGERRQLIVP PHLAHGESGA RGVPGSAVLL FEVELVSRED Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
MAF acts as a transcriptional activator or repressor. The protein is involved in embryonic lens fiber cell development.Specifications
FKBP10 | |
Polyclonal | |
Unconjugated | |
FKBP10 | |
65 kDa FK506-binding protein; 65 kDa FKBP; AI325255; BRKS1; FK506 binding protein 10; FK506 binding protein 10, 65 kDa; FK506 binding protein 6 (65 kDa); FK506-binding protein 10; FKBP10; FKBP-10; Fkbp1-rs; Fkbp6; FKBP65; FKBP-65; Fkbprp; Fkbp-rs; Fkbp-rs1; hFKBP65; Immunophilin FKBP65; OI11; OI6; Peptidyl-prolyl cis-trans isomerase FKBP10; PPIASE; PPIase FKBP10; PSEC0056; Rotamase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60681 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96AY3 | |
FKBP10 | |
Recombinant protein corresponding to Human FKBP10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only