Learn More
FKBP5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529735
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32, MCF-7. Predicted reactivity: Mouse (92%), Rat (96%), Pig (98%), Chicken (88%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FKBP5 is a member of the immunophilin protein family. This family plays a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP5 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It is thought to mediate calcineurin inhibition. It also interacts functionally with mature hetero-oligomeric progesterone receptor complexes along with the 90 kDa heat shock protein and P23 protein. The gene that encodes FKBP5 has been found to have multiple polyadenylation sites. Alternative splicing results in multiple transcript variants.Specifications
FKBP5 | |
Polyclonal | |
Unconjugated | |
FKBP5 | |
51 kDa FK506-binding protein; 51 kDa FKBP; 54 kDa progesterone receptor-associated immunophilin; AIG6; androgen-regulated protein 6; D17Ertd592e; Dit1; EC 5.2.1.8; FF1 antigen; FK506 binding protein 5; FK506-binding protein 5; FKBP prolyl isomerase 5; Fkbp5; FKBP-5; FKBP51; FKBP-51; FKBP54; HSP90-binding immunophilin; p54; peptidylprolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase FKBP5; PPIase; PPIase FKBP5; Ptg-10; rotamase; T-cell FK506-binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2289 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q13451 | |
FKBP5 | |
Recombinant fragment corresponding to a region within amino acids 220 and 457 of Human FKBP5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |