Learn More
FKBPL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560378
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TVLHANLAAC QLLLGQPQLA AQSCDRVLER EPGHLKALYR RGVAQAALGN LEKATADLKK VLAIDPKNRA AQEELGKVVI QGKNQDAGLA Q Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 91%.
The protein encoded by this gene has similarity to the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The encoded protein is thought to have a potential role in the induced radioresistance. Also it appears to have some involvement in the control of the cell cycle.Specifications
FKBPL | |
Polyclonal | |
Unconjugated | |
Fkbpl | |
DIR1; FK506 binding protein like; FK506 binding protein-like; FK506-binding protein like; FK506-binding protein-like; FKBP4; FKBPL; immunophilin-like protein NG7; NG7; peptidyl-prolyl cis-trans isomerase; Ppiase-X; WAF-1/CIP1 stabilizing protein 39; WISp39 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
63943 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UIM3 | |
Fkbpl | |
Recombinant protein corresponding to Human FKBPL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only