Learn More
Flash Point Standard - Cleveland Open Cup, ASTM D 92 (nominal value: 148 °C), Paragon Scientific™
Specifically designed for use in regular instrument performance checks. Packed uniquely in 3 x 80L single shot bottles assuring the highest level of sample integrity. Typically used in the analysis of petroleum products with flash points above 79°C (175°F) and below 400°C (752°F)
Brand: Paragon Scientific FP-COC-3
Additional Details : Weight : 1.00000kg
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Packaged in 3 X 80mL bottles
- One bottle per test ensures zero contamination / high end volatile loss
- Fully traceable to International Standards
- Supplied with MSDS
- Tamper evident security packaging
- 12 Month shelf life
- Manufactured in the United Kingdom
Specifications
3 x 80 mL |
Safety and Handling
- X3 80ML FP-COC-3 Cleveland Open Cup Flash Point St
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H332-Harmful if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P331-Do NOT induce vomiting.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Dec-1-ene, dimers, hydrogenated
Nominal values quoted. Certified Values will vary from batch to batch.