missing translation for 'onlineSavingsMsg'
Learn More

Flash Point Standard - Cleveland Open Cup, ASTM D 92 (nominal value: 188 °C), Paragon Scientific™

Specifically designed for use in regular instrument performance checks. Packed uniquely in 3 x 80L single shot bottles assuring the highest level of sample integrity. Typically used in the analysis of petroleum products with flash points above 79°C (175°F) and below 400°C (752°F)

Brand:  Paragon Scientific FP-COC-4


Product Code. 16030232

  • £170.00 / Pack of 3

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Packaged in 3 X 80mL bottles
  • One bottle per test ensures zero contamination / high end volatile loss
  • Fully traceable to International Standards
  • Supplied with MSDS
  • Tamper evident security packaging
  • 12 Month shelf life
  • Manufactured in the United Kingdom
Specifications

Specifications

3 x 80 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X3 80ML FP-COC-4 Cleveland Open Cup Flash Point St

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted. Certified Values will vary from batch to batch.