Learn More
Flash Point Standard, COC ASTM D92 (Nominal Value: 262.0 °C ), Paragon Scientific™
These are certified reference materials characterized by a method specific multi-laboratory study in accordance with ASTM D92. These materials are suitable for verifying the performance of an instrument and/or procedure as specified in this standard.
Brand: Paragon Scientific CRMU-COC-HIGH
Additional Details : Weight : 1.00000kg
Description
- Certified Reference Material for annual verification checks in accordance with ASTM D92.
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
- Packaged in 3 x 80 mL bottles – ‘One shot’, one bottle per test ensures zero contamination/ high-end volatile loss.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
3 x 80 mL |
Nominal values quoted. Certified Values will vary from batch to batch.