Learn More
Flash Point Standard - Pensky Martens, ASTM D 93 Proc. A (nominal value: 116.0 °C), Paragon Scientific™
UKAS certified standards are specifically designed for use in regular instrument performance checks. Packed in 3 x 80mL single shot bottles assuring the highest level of sample integrity. Typically used in the analysis of petroleum products with flash points above 40°C and below 360°C.
Brand: Paragon Scientific FP-PMCC-3
Additional Details : Weight : 1.00000kg
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Packaged in 3 X 80mL bottles
- One bottle per test ensures zero contamination / high end volatile loss
- Fully traceable to International Standards
- Supplied with MSDS
- Tamper evident security packaging
- 12 Month shelf life
- Manufactured in the United Kingdom
Specifications
3 x 80 mL |
Safety and Handling
- Flash Point Standard - Pensky Martens
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : kerosine
Nominal values quoted. Certified Values will vary from batch to batch.