Learn More
Flash Point Standard - Pensky Martens, ASTM D 93 Proc. A (nominal value: 59.0 °C), Paragon Scientific™
UKAS certified standards are specifically designed for use in regular instrument performance checks. Packed in 3 x 80mL single shot bottles assuring the highest level of sample integrity. Typically used in the analysis of petroleum products with flash points above 40°C and below 360°C.
Brand: Paragon Scientific FP-PMCC-1
Additional Details : CAS Number : 68334-30-5 Weight : 1.00000kg Transport : UN number : 1202 Chem class : 3 Pack group : III
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Packaged in 3 X 80mL bottles
- One bottle per test ensures zero contamination / high end volatile loss
- Fully traceable to International Standards
- Supplied with MSDS
- Tamper evident security packaging
- 12 Month shelf life
- Manufactured in the United Kingdom
Specifications
3 x 80 mL |
Safety and Handling
- flash point standard - Pensky Martens
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : diesel fuel
Nominal values quoted. Certified Values will vary from batch to batch.