Learn More
Flash Point Standard, PMCC ASTM D 93 Proc A (nominal value 77.5 °C), Paragon Scientific™
These are certified reference materials characterized by a method specific multi-laboratory study in accordance with ASTM D93 Procedure A. These materials are suitable for verifying the performance of an instrument and/or procedure as specified in this standard
Brand: Paragon Scientific CRMU-PMCC-LOW
Additional Details : Weight : 1.00000kg
Description
- Certified Reference Material for annual verification checks in accordance with ASTM D93, Procedure A.
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
- Packaged in 3 x 80 mL bottles – ‘One shot’, one bottle per test ensures zero contamination/ high-end volatile loss.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom.
Specifications
3 x 80 mL |
Safety and Handling
- X3 80ML CRM-PMCC-Low Flash Point Standard
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Distillates (petroleum), hydrotreated light
Nominal values quoted. Certified Values will vary from batch to batch.