Learn More
Gas Arc™ Inline Flashback Arrestor
£104.37 - £178.00
Product Code | Brand | For Use With (Application) | Pressure | Connection Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Pressure | Connection Type | Price | Quantity & Availability | |||
12960820
|
Gas Arc™
FBA07048 |
Fuel Gas | 5 bar (inlet)/5 bar (outlet) | 3/8 BSP LH Female (inlet); 3/8 BSP LH Male (outlet) |
£104.37
Each |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12950820
|
Gas Arc™
FBA07051 |
Oxygen | 20 bar (inlet)/20 bar (outlet) | 3/8 BSP RH Female (inlet); 3/8 BSP RH Male (outlet) |
£178.00
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Specifications
Inline Flashback Arrestor |