missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ FlashGel™ System DNA Cassettes

Cassettes optimized for DNA migration. Lonza™ FlashGel™ System DNA Cassettes separate DNA quickly and DNA migration can be seen as it happens. Separates DNA in 2 to 7 minutes.

Brand:  Lonza™ 57032


 View more versions of this product

Product Code. 11507865

  • £313.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Get results in 5 minutes or less
  • Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2.2%
10 to 1000 bp
2.2% Agarose
Agarose Gel, Stain and Buffer
9 Pack
4.5 in.
11.5 cm
4.2 in.
8.4 cm
1.7 cm
DNA Cassettes
16 + 1-well
Electrophoresis
Agarose
18° to 26°C
2.7 in.
7 cm
10.7 cm
0.6 in.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers