Learn More
BUCHI FlashPure Chromatography Solid Loader
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
POA
Product Code | Brand | Size (Metric) Tube Rack | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) Tube Rack | Price | Quantity & Availability | |||||
16491374
|
BUCHI
11068975 |
15 g |
POA
1 set |
N/A | |||||
Description
- Screw cap makes sealing easy
- Various sizes for differing sample amounts
- Choose a size large enough for the sample
- Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BUCHI FlashPure Chromatography Solid Loader