missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ Flask Carrier without Handle
Designed with a round bottom to safely hold round-bottomed flasks.
Brand: IKA™ 0025003280
Additional Details : Weight : 0.25000kg
Description
- Round bottom
- Anodized aluminum construction
- Temperature resistant
Specifications
250 mL | |
Up to 300°C | |
142 mm |
Aluminum | |
Round |