Learn More
Thermo Scientific™ Flat Caps for UTW PCR Plates, strips of 8, optical clear
Minimize vapor loss with these flat cap strips for use with Thermo Scientific™ Piko™ 24-well plates in the Piko Thermal Cycler and the PikoReal Real-Time PCR System.
Brand: Thermo Scientific™ TCS1080
Code : A0
Additional Details : Weight : 1.87330kg
Description
All our sealing products are designed to provide the ultimate protection while also being easy to use. Thermo Scientific qPCR sealing options are optically clear to deliver maximum and consistent signal transmission, critical for accurate qPCR results.
These flat cap strips provide an extremely tight seal in UTW (ultra-thin walled) PCR plates. They are an ideal choice for low volume reactions where it is important to minimize vapor loss. In addition, our optically-clear flat caps provide great transmission for all fluorescence detection applications, such as qPCR.
Highlights
- 8-cap strip
- Optically clear for fluorescence detection
- Tight sealing for low-volume samples and consistent quantification results
- Ideal for sealing 24-well Piko UTW PCR plates
Specifications
Clear | |
8 | |
PCR Plate Cap Strip | |
Yes |
Polypropylene | |
Strips of 8 Optical Caps | |
Piko | |
96-well Plate |