missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck and One Angled Side Neck
£146.00 - £554.00
Specifications
Material | Borosilicate Glass, Duran Glass |
---|---|
For Use With (Application) | Reaction vessels |
Product Type | Flange lid |
Product Code | Brand | Closure Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Includes | Price | Quantity & Availability | ||||
11572344
|
Lenz Laborglasinstrumente™
06102214 |
Center Neck (NS): 29/32 & Side Neck: 14/23 | Laboratory Flange 100 |
£203.00
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
10236409
|
Lenz Laborglasinstrumente™
06102244 |
Center Neck (NS): 29/32, Side Neck: 29/32 | Laboratory Flange 100 |
£215.00
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11582344
|
Lenz Laborglasinstrumente™
06102264 |
Center Neck (NS): 45/40 | Laboratory Flange 100 |
£239.00
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11512354
|
Lenz Laborglasinstrumente™
06102544 |
Center Neck (NS): 29/32 | Laboratory Flange 200 |
£533.00
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11502354
|
Lenz Laborglasinstrumente™
06102464 |
Center Neck (NS): 45/40 | Laboratory Flange 150 |
£356.00
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11592344
|
Lenz Laborglasinstrumente™
06102444 |
Center Neck (NS): 29/32 | Laboratory Flange 150 |
£335.00
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11522354
|
Lenz Laborglasinstrumente™
06102564 |
Center Neck (NS): 45/40 | Laboratory Flange 200 |
£554.00
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11552344
|
Lenz Laborglasinstrumente™
06102111 |
Center Neck (NS): 14/23 | Laboratory Flange 60 |
£146.00
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11562344
|
Lenz Laborglasinstrumente™
06102141 |
Center Neck (NS): 29/32 | Laboratory Flange 60 |
£155.00
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
Specifications
Borosilicate Glass, Duran Glass | |
Reaction vessels | |
Flange lid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title