Learn More
SCAT™ Flat Flange Lids for Glass Reactors
The flat flange lid is usually placed between two flanges to be joined and then tightened with bolts to create a secure connection. Suitable for glass reactors with a SCHOTT™ flat flange.
Brand:  SCAT™ 130100
153.39 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17839056
  • £178.00 / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Flat flange allows the lid to be securely attached to the reactor body and ensures a reliable seal to prevent leakage
  • Compatible according to DIN 12214 (laboratory flat flange) and compatible with glass reactors for seamless integration
  • Effortless and hassle-free to assemble, saving time and effort
  • Flexibly expand our products to fit your specific needs and meet future needs
  • Made entirely of PTFE, for universal chemical resistance to a wide range of harsh chemicals, increasing application versatility and reliability
  • Equipped with practical attachments that allow easy connection and use of existing equipment that has ground or GL threads
  • Important component for the safe and reliable connection of pipes and vessels in various industrial applications to prevent leaks and keep equipment running smoothly
Specifications
Specifications
Flat Flange
Glass Reactors
DN 100
Connection of Pipes and Vessels, Preventing Leaks
1 x NPT 1/2 in. (f) Centered, 2 x NPT 1/2 in. (f) Directly in Center, 1 x NPT 1/2 in. (f) Beside Center
PTFE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products