Learn More
Thermo Scientific™ Flat PCR Cap Strips Promotion
Optimize PCR applications with these flat cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
Brand: Thermo Scientific™ AB0784
Additional Details : Weight : 0.17000kg
Description
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Specifications
Clear Frosted | |
Virgin Medical Grade Polypropylene | |
250 | |
250/Cs. | |
Strips of 8 Flat Caps |
Certified free of human DNA, RNase, DNase, and endotoxins. | |
96 | |
250 | |
Facilitates sample labeling | |
96-well PCR Plate, Standard and Low-profile PCR Tube Strips |
0