Learn More
Thermo Scientific™ Flat PCR Caps, strips of 12
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
Brand: Thermo Scientific™ AB0850
Code : 03
Additional Details : Weight : 1.00000kg
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Highlights
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Specifications
Natural | |
Polypropylene | |
Thermal Cycler |
Certified DNA/RNase Free | |
80 Strip Caps | |
Flat Cap, 12-Cap Strip |