Learn More
Memmert™ Flat Covers for Water Bath
Flat stainless steel cover with concentric ring sets for ring set openings: 6 openings / 87 mm diameter
Brand: Memmert™ B02682
Code : 81
Additional Details : Weight : 0.10000kg
Specifications
Stainless Steel | |
6 | |
3 Years |