Learn More
3M™ Petrifilm™ Flat Spreader 6425
Spreader used to distribute the sample evenly for 3M™ Petrifilm™ Staph Express Count Plates.
Brand: 3M™ Petrifilm™ 7100002605
53.09 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
- Flat spreader for 3M™ Petrifilm™ Staph Express Count Plates.
- Spreads inoculum over uniform area.
- Clear acrylic material allows the technician to observe and control spread.
Specifications
Accessory | |
2 Pk. | |
3 M™ Petrifilm™ Staph Express Count Plates. |