missing translation for 'onlineSavingsMsg'
Learn More
Flavin adenine dinucleotide disodium salt hydrate, 94% (dry wt.), water <10%, Thermo Scientific Chemicals Product Code.: 11411838

Flavin adenine dinucleotide disodium salt hydrate, 94% (dry wt.), water <10%, Thermo Scientific Chemicals

Product Code. 11411838
100 mg
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy

Product Code. 11411838

Brand: Thermo Scientific Alfa Aesar A14495.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.

Solubility
Soluble in water (50 mg/ml).

Notes
Store at 4°C. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1891059-93-0
Molecular Formula C27H31N9Na2O15P2
Molecular Weight (g/mol) 829.52
MDL Number MFCD00151217
InChI Key XLRHXNIVIZZOON-SJFBGTSINA-L
Synonym flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5'-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tnShow MoreShow Less
PubChem CID 131675332
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrateShow MoreShow Less
SMILES [Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1Show MoreShow Less

Specifications

Quantity 100 mg
Solubility Information Soluble in water (50mg/ml).
Formula Weight 829.52
Percent Purity 94%
Assay (dry wt.), water <10%
Chemical Name or Material Flavin adenine dinucleotide disodium salt hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250532 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog NumberA14495.MC,A14495.ME
Lot Number10248223 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog NumberA14495.MC,A14495.ME
Lot Number10239443 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14495.MC,A14495.ME
Lot Number10237752 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14495.MC,A14495.ME
Lot Number10164650 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14495.MC,A14495.ME

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Flavin adenine dinucleotide disodium salt hydrate, 94% (dry wt.), water <10%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.