missing translation for 'onlineSavingsMsg'
Learn More

Flavone, 99+%, Thermo Scientific Chemicals

Product Code. 10122340
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 525-82-6
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.24
InChI Key VHBFFQKBGNRLFZ-UHFFFAOYSA-N
Synonym flavone, 2-phenylchromone, 2-phenyl-4h-chromen-4-one, 2-phenyl-4-chromone, 2-phenyl-4-benzopyron, 2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2-phenyl, asmacoril, chromocor, cromaril
PubChem CID 10680
ChEBI CHEBI:42491
IUPAC Name 2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2

Specifications

Melting Point 95.0°C to 98.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 373
Merck Index 15, 4124
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in most common organic solvents
Formula Weight 222.24
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Flavone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'