missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flavone, 99+%, Thermo Scientific Chemicals
£18.70 - £173.00
Chemical Identifiers
CAS | 525-82-6 |
---|---|
Molecular Formula | C15H10O2 |
Molecular Weight (g/mol) | 222.24 |
InChI Key | VHBFFQKBGNRLFZ-UHFFFAOYSA-N |
Synonym | flavone, 2-phenylchromone, 2-phenyl-4h-chromen-4-one, 2-phenyl-4-chromone, 2-phenyl-4-benzopyron, 2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2-phenyl, asmacoril, chromocor, cromaril |
PubChem CID | 10680 |
ChEBI | CHEBI:42491 |
IUPAC Name | 2-phenylchromen-4-one |
SMILES | C1=CC=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10050930
|
Thermo Scientific Acros
119160010 |
1 g | Glass bottle |
£18.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10122340
|
Thermo Scientific Acros
119160050 |
5 g | Glass bottle |
£48.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10056403
|
Thermo Scientific Acros
119160250 |
25 g | Glass bottle |
£173.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
525-82-6 | |
222.24 | |
flavone, 2-phenylchromone, 2-phenyl-4h-chromen-4-one, 2-phenyl-4-chromone, 2-phenyl-4-benzopyron, 2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2-phenyl, asmacoril, chromocor, cromaril | |
CHEBI:42491 | |
C1=CC=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2 |
C15H10O2 | |
VHBFFQKBGNRLFZ-UHFFFAOYSA-N | |
10680 | |
2-phenylchromen-4-one |
Specifications
525-82-6 | |
White | |
99% min. (HPLC) | |
17, 373 | |
flavone, 2-phenylchromone, 2-phenyl-4h-chromen-4-one, 2-phenyl-4-chromone, 2-phenyl-4-benzopyron, 2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2-phenyl, asmacoril, chromocor, cromaril | |
VHBFFQKBGNRLFZ-UHFFFAOYSA-N | |
2-phenylchromen-4-one | |
10680 | |
222.24 | |
Crystalline Powder |
95.0°C to 98.0°C | |
Authentic | |
C15H10O2 | |
15, 4124 | |
Solubility in water: practically insoluble in water. Other solubilities: soluble in most common organic solvents | |
C1=CC=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2 | |
222.24 | |
CHEBI:42491 | |
99+% | |
Flavone |
Safety and Handling
missing translation for 'einecsNumber' : 208-383-8
RUO – Research Use Only