Learn More
OHAUS™ Flex System Clamp
OHAUS™ Flex System Clamp is designed to securely grip and position a variety of laboratory glassware.
Brand:  OHAUS™ 30400011
Additional Details : Weight : 2.30000kg
 View more versions of this product
Product Code. 15732429
  • £263.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Corrosion-resistant stainless steel clamps provide excellent chemical resistance, durability, and can be autoclaved. Nickel-plated zinc clamps offer high-tensile strength at an economical price.
  • Prevent accidents in the lab! Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects. Select from a variety of grip sizes, prong styles and adjustment types.
  • Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
Specifications
Specifications
Flex System Clamp
Clamp, Specialty, Flex, CLS-FLEXSM
13.000 mm
5.00 in.
127 mm
5.00 in.
Nickel-plated
13 mm
0.50 in.
12.000 in.
305 mm
127.00 mm